missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ 47 mm Microfiltration Glassware Replacement Parts

Replacement Parts for Corning PYREX 47mm Microfiltration Glassware Apparatus (13-645-088, sold separately)

POA

Specifications

For Use With (Equipment) PYREX 47 mm Microfiltration Glassware Apparatus
No. per Case 1
No. per Pack 1/Pk.
View More Specs

Description

Description

Replacement Parts for Corning™ PYREX™ 47 mm Microfiltration Glassware Apparatus (13-645-088, sold separately).
Specifications

Specifications

PYREX 47 mm Microfiltration Glassware Apparatus
1
1/Pk.
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.