missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring

Catalog Number p-8002560
79.46 EUR valid until 2024-12-31
Use promo code "21932" to get your promotional price.


Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C
Closure Material Polypropylene
Closure Size 25 mm
Closure Type Screw Cap
Diameter (Metric) 36 mm
For Use With (Equipment) Ideal for Mixing and Storage of Culture Medium
Height (English) 2.8 in.
Height (Metric) 70 mm
Product Type Reagent Bottle
Capacity (Metric) 25 mL
Capacity (English) 0.8 oz.
Color Clear
Diameter (English) 1.4 in.
For Use With (Application) Ideal for mixing and storage of culture medium
Includes Polypropylene screw cap and pouring ring
Material Borosilicate Glass
No. per Pack 10/PK
Quantity 10 Pk.
Thread Size (Metric) GL 25
Certifications/Compliance ISO 4796
Closure Included Yes
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Wide
Shape Round
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'