Learn More
Corning™ CoolRack™ XT CFT24, Holds 24 cryovial or FACS tubes
Holds 24 cryogenic vial or FACS tubes, with gripping wells for one-hand vial opening/closing.
Brand: Corning™ 432050
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensure +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- SBS-compatible
- "Gripping" wells for one-hand vial opening/closing
Specifications
24 Cryogenic vial or FACS Tubes | |
CFT Tube Modules |