Learn More
PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
With glass pennyhead stopper
Brand:  PYREX™ 34505-36
Code : A0
Additional Details : Weight : 0.80000kg
Product Code. 11760269
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for standard taper glass stoppers. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 1 mL and 2 mL sizes are test tube shaped. Sizes 3L through 6L have pennyhead standard taper stoppers.
For stopper only, see Cat. Nos. 7650 and 7661.