missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Gooch Crucible with Integral Glass Sintered Disc

Ideal for drying precipitates to constant mass at 110°C. Pyrex™ Gooch Crucible includes an integral sintered disc.

Brand:  Pyrex 3660/06M

Code : 33

Additional Details : Weight : 0.24628kg

 View more versions of this product

Product Code. 11961548

Quantity Price
1 104.80 € / Pack of 5
10 96.61 € / Pack of 5
Estimated Shipment: 06-07-2021

Add to basket


Description

Description

Ideal for drying precipitates to constant mass at 110°C. PyrexTM Gooch Crucible includes an integral sintered disc.

  • Ideal for drying precipitates to constant mass at 110°C
  • Also suitable for ignition work up to 450°C (see safety notice)
  • With integral glass sintered disc
  • Manufactured from chemically resistant Pyrex™ borosilicate glass
Specifications

Specifications

With Integral Glass sintered disc
Ideal for drying precipitates to constant mass at 110°C and for ignition work up to 450°C
30mL
Pyrex Borosilicate Glass
Gooch crucible
30mm
450°C
53mm
3
Videos
SDS
Documents
Certificates