missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Straight Bore Repair Burette
Straight bore burette.
Brand: Pyrex SST502
Code : NEW
Informations supplémentaires : Weight : 0.52000kg
Description
- Straight bore burette
Spécification
Buret | |
1.5 mm, Top: 12 mm, Stopcock: 12.5 mm | |
Borosilicate Glass | |
8 mm |