missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals
19.80 EUR valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 205.0°C |
Color | Orange |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic bottle |
Quantity | 25 g |
Fieser | 06,498; 07,308; 08,425; 09,397; 10,334; 11,450; 12,256; 13,237; 14,269; 15,57 |
Merck Index | 15, 8086 |
Solubility Information | Solubility in water: .. Other solubilities: insoluble in dichloromethane |
Formula Weight | 215.56 |
Show More |
Safety and Handling
Product Identifier
- Pyridinium chlorochromate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H272-May intensify fire; oxidiser.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'