missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals

Catalog Number 10012270
39.30 EUR valid until 2024-07-31
Use promo code "23251" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
26299-14-9
C5H6ClCrNO3
215.56
MFCD00013106
PCC
26299-14-9
C5H6ClCrNO3
215.56
MFCD00013106
PCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 205.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Fieser 06,498; 07,308; 08,425; 09,397; 10,334; 11,450; 12,256; 13,237; 14,269; 15,57
Merck Index 15, 8086
Solubility Information Solubility in water: .. Other solubilities: insoluble in dichloromethane
Formula Weight 215.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium chlorochromate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyridinium chlorochromate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Oxidising solid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'