Learn More
Pyrimidine, 99%, ACROS Organics™
Brand: Acros Organics 157700050
Code : 29
Additional Details : CAS Number : 289-95-2 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
289-95-2 | |
80.09 | |
CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
9260 | |
pyrimidine |
C4H4N2 | |
MFCD00006059 | |
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
CHEBI:16898 | |
C1=CN=CN=C1 |
Specifications
Pyrimidine | |
31°C | |
Glass bottle | |
1.01 | |
Colorless to Orange | |
5g | |
98.5% min. (GC) | |
MFCD00006059 | |
15, 8100 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
C1=CN=CN=C1 | |
80.09 | |
CHEBI:16898 | |
Liquid |
1.0100g/mL | |
Authentic | |
1.5010 to 1.504 | |
123°C to 124°C | |
20°C to 22°C | |
289-95-2 | |
C4H4N2 | |
23, 89 | |
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l | |
CZPWVGJYEJSRLH-UHFFFAOYSA-N | |
pyrimidine | |
9260 | |
80.09 | |
99% |
Safety and Handling
- Pyrimidine
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.