missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxylic acid hydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-4510706
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
336100-46-0
C5H5NO2
111.10
MFCD06201862
DOYOPBSXEIZLRE-UHFFFAOYSA-N
pyrrole-3-carboxylic acid hydrate, 1h-pyrrole-3-carboxylic acid hydrate, c5h5no2.h2o, 1h-pyrrole-3-carboxylicacid, hydrate 1:?
45076181
1H-pyrrole-3-carboxylic acid;hydrate
OC(=O)C1=CNC=C1
This item is not returnable. View return policy
336100-46-0
C5H5NO2
111.10
MFCD06201862
DOYOPBSXEIZLRE-UHFFFAOYSA-N
pyrrole-3-carboxylic acid hydrate, 1h-pyrrole-3-carboxylic acid hydrate, c5h5no2.h2o, 1h-pyrrole-3-carboxylicacid, hydrate 1:?
45076181
1H-pyrrole-3-carboxylic acid;hydrate
OC(=O)C1=CNC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 144.0°C to 147.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 22,II,17
Solubility Information Solubility in water: soluble. Other solubilities: soluble in benzene, diethyl ether, petrol. ether
Formula Weight 111.1
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Pyrrole-3-carboxylic acid hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'