missing translation for 'onlineSavingsMsg'
Learn More

Pyruvonitrile, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7057206
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
631-57-2
C3H3NO
69.063
MFCD00001838
QLDHWVVRQCGZLE-UHFFFAOYSA-N
pyruvonitrile, propanenitrile, 2-oxo, oxopropionitrile, 2-oxopropiononitrile, 2-oxopropionitrile, oxypropionitrile, pyruvic acid nitrile, unii-ulo441649v, accn, .alpha.-oxopropionitrile
69430
acetyl cyanide
CC(=O)C#N
This item is not returnable. View return policy
631-57-2
C3H3NO
69.063
MFCD00001838
QLDHWVVRQCGZLE-UHFFFAOYSA-N
pyruvonitrile, propanenitrile, 2-oxo, oxopropionitrile, 2-oxopropiononitrile, 2-oxopropionitrile, oxypropionitrile, pyruvic acid nitrile, unii-ulo441649v, accn, .alpha.-oxopropionitrile
69430
acetyl cyanide
CC(=O)C#N

Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

Solubility
Soluble in ether and acetonitrile.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Brown
Density 0.974 g/mL
Boiling Point 92°C to 93°C
Flash Point 14°C (57°F)
Linear Formula CH3COCN
Refractive Index 1.3755
Quantity 1 g
UN Number UN3273
Beilstein 1737633
Solubility Information Soluble in ether and acetonitrile.
Formula Weight 69.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Pyruvonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'