missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 with 2 Doors
Safe and approved storage of flammable hazardous substances in working areas
2246.14 € - 3331.05 €
Specifications
Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 620 mm |
Depth (Metric) Interior | 519 mm |
Dimensions (D x W x H) | 620 x 1164 x 1947 mm |
Dimensions (D x W x H) Interior | 519 x 1050 x 1626 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Description | Weight (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Description | Weight (Metric) | Price | Quantity & Availability | |||
15546895
|
asecos™
3100306033635 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Body: Antharcite grey, Door: Warning yellow RAL1004; Sheet Steel RAL 7035 | 271 kg |
2246.14 €
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15986486
|
asecos™
3100306133635 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Body: Antharcite grey, Door: Light grey RAL 7035; Sheet Steel RAL 7035 | 271.50 kg |
2246.14 €
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966496
|
asecos™
3100306133647 |
4 x drawer (sheet steel powder-coated) | Body: Antharcite grey, Door: Light grey RAL 7035; Sheet Steel RAL 7035 | 302 kg |
2887.97 €
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15966466
|
asecos™
3100306033647 |
4 x drawer (sheet steel powder-coated) | Body: Antharcite grey, Door: Warning yellow RAL1004; Sheet Steel RAL 7035 | 302 kg |
2887.97 €
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15986496
|
asecos™
3100306133649 |
6 x drawer (sheet steel powder-coated) | Body: Antharcite grey, Door: Light grey RAL 7035; Sheet Steel RAL 7035 | 334 kg |
3331.05 €
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Warranty
10 years
Specifications
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
519 mm | |
519 x 1050 x 1626 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1626 mm | |
2 | |
1050 mm |
620 mm | |
620 x 1164 x 1947 mm | |
Steel | |
Wing | |
1947 mm | |
Q30.195.116 | |
1164 mm |