missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Qiagen™ Robotic Tips
GREENER_CHOICE

Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series

229.48 € - 381.00 €

Specifications

Certifications/Compliance Non-ANSI
Clean Claims DNase, Rnase Free
Format Rack
Tip Style Automation, Round
For Use With (Equipment) Qiagen BioRobot
View More Specs

Products 3
Product Code Brand Color Sterility Volume (Metric) Price Quantity & Availability  
Product Code Brand Color Sterility Volume (Metric) Price Quantity & Availability  
12545467
View Documents Promotion Details
Axygen™
QT-1100-CBK-R
Black Non-sterile 1100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13417699
GREENER_CHOICE
View Documents Promotion Details
Axygen™
QTF-1100-CBK-R-S
Black Sterile 1000 μL
376.00 €
Pack of 1536
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
12565467
GREENER_CHOICE
View Documents Promotion Details
Axygen™
QT-300-CBK-R
Black Non-sterile 300 μL
381.00 €
Pack of 2304
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

Available in volumes of 250, 300, 1000 and 1100 μL.

  • Clear and conductive black tips available
  • Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips

Specifications

Specifications

Non-ANSI
Rack
Qiagen BioRobot
DNase, Rnase Free
Automation, Round
Videos
SDS
Documents

Documents

Product Certifications