Learn More
Socorex™ Qualitix™ Micropipette Tips, 200μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.
29.60 € - 110.00 €
Specifications
Volume (Metric) | 200 μL |
---|---|
Volume (Metric) Dispensing | 200μL |
Autoclavable | Autoclavable |
Disposable | Single-use |
Product Line | Qualitix |
Product Code | Brand | Packaging | Sterility | Tip Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Sterility | Tip Style | Price | Quantity & Availability | |||
15700649
|
Socorex™
3070.0200B |
1000/Bag | Non-sterile | Beveled |
29.60 €
Pack of 1000 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
15740649
|
Socorex™
3070.0200R |
96/Rack | Non-sterile | Beveled |
69.80 €
Pack of 960 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
15750649
|
Socorex™
3070.0200RS |
96/Rack | Sterile | Beveled |
79.30 €
Pack of 960 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||
15760679
|
Socorex™
3290.0200B |
1000/Bag | Non-sterile | Regular Tip, Bevel Point |
29.60 €
Pack of 1000 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
15770679
|
Socorex™
3290.0200R |
96/Rack | Non-sterile | Regular Tip, Bevel Point |
69.80 €
Pack of 960 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
15780659
|
Socorex™
3080.0200GR |
96/Rack | Non-sterile | Gel Loading, Capillary |
110.00 €
Pack of 576 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
15780679
|
Socorex™
3290.0200RS |
96/Rack | Sterile | Regular Tip, Bevel Point |
79.30 €
Pack of 960 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||
Description
- Cover a range of sizes.
- Autoclavable.
- Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
- Various tip versions available.
Specifications
200 μL | |
Autoclavable | |
Qualitix |
200μL | |
Single-use |