Learn More
Macherey-Nagel™ Quantofix™ Arsenic 10 Semi-quantitative Test Strips
Allows quick and easy determination of arsenic in solutions. Macherey-Nagel™ Quantofix™ Arsenic 10 Semi-quantitative Test Strips comes with all necessary reagents so that measurements can be started immediately, with reliable results available in just 30 minutes.
Brand: Macherey-Nagel™ 91334
Code : 26
Additional Details : CAS Number : 56-37-1 Weight : 0.08000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
As a naturally occurring element, arsenic is widely distributed in the Earth’s crust. In the natural environment it is most frequently present as inorganic arsenic compound in combination with sulphur or oxygen. Organic arsenic compounds can be used as pesticides. Arsenic is toxic and causes skin diseases, keratosis and melanomas.
- Arsenic levels in drinking water have to be monitored thoroughly
- Quantofix™ can reliably monitor the WHO threshold value of 0.01mg/L
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
White to Yellow/Brown | |
Determination of Arsenic 10 | |
Test strips, aluminum strip case with color chart, reagents in drop/measuring spoon caps, reaction vessel, syringe, needle | |
15°C to 30°C | |
Semi-qualitative Test Strip |
Strip | |
0, 0.01, 0.025, 0.05, 0.1, 0.5mg/L As3+/5+ | |
100 Pack | |
30 min. |
Safety and Handling
- QUANTOFIX Arsenic 10
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- H411-Toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : potassium permanganate, aminosulfonic acid (sulfamic acid),zinc powder (stabilizied)