Learn More
Macherey-Nagel™ Quantofix™ Chlorine Test Strip Kits
Allows for the quick and easy determination of chlorine in solutions. Macharey Nagel™ Quantofix™ Chlorine Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand: Macherey-Nagel™ 91317
Code : 26
Additional Details : Weight : 0.17100kg
Description
- Ready to use
- Rapid: provides reliable results within one minute
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
White to Red/Violet | |
Determination of Chlorine | |
100 Pack | |
1 min. |
Strip | |
0, 1, 3, 10, 30, 100mg/L Cl2 | |
30°C | |
Semi-qualitative Test Strip |
Safety and Handling
- QUANTOFIX Chlorine
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : pyridine, potassiumcyanide