Learn More
Thermo Scientific Chemicals Quartz Discs
Manufactured by fusing naturally occurring crystalline silica
Brand:  Thermo Scientific Chemicals 042299.KS
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15446455
  • 57.70 € / Each
In Stock
to see stock.
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Mineral, Quartz Disc
Ground and polished
50.8 mm
3.18 mm