missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Quartz Suprasil Fluorescence Cells

Brand:  Hellma™ 109004F-10-40

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11786234

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Fluorescence Cell
Quartz Suprasil
2/Pack
45 mm
Magnetic Stirrers
1500 μL
4mm
10 x 4 mm
12.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers