missing translation for 'onlineSavingsMsg'
Learn More

Quercetin hydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-190229
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
849061-97-8
C15H10O7
302.24
MFCD03847906
REFJWTPEDVJJIY-UHFFFAOYSA-N
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
16212154
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1
This item is not returnable. View return policy
849061-97-8
C15H10O7
302.24
MFCD03847906
REFJWTPEDVJJIY-UHFFFAOYSA-N
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
16212154
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 18, 242
Merck Index 14, 8034
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,acetone,pyridine,,glacial acetic acid
Formula Weight 302.24
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Quercetin hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Quercetin hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'