Learn More
SCAT Quick-Lock Connectors
Quick-Lock Connectors provide easy and safe connection of capillary tubing and SCAT Safety Caps. Connect and disconnect with a “click” - without dripping or air bubbles. The valve closes automatically when disconnected. Included PFA fittings allow all common hose diameters to be connected.
Brand:  SCAT 360180
36.30 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.08600kg
 View more versions of this product
Product Code. 17332840
  • 45.95 € / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
The Quick-Lock Connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
  • Made of chemically resistant glass-filled polypropylene, PEEK™ materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1.6 (1/16 in.) / 2.3 /3.2 (1/8 in.) mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1.0 bar pressure resistance
Specifications
Specifications
1 x PFA-ferrule 1.6/2.3/3.2 mm ID
HPLC Systems
Quick-Lock Connector (m) for 1 Safety Cap
Capillaries, Tubes and Safety Caps