Learn More
Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
75.40 € - 610.00 €
Specifications
Certifications/Compliance | ISO 4797 |
---|---|
Material | Borosilicate Glass |
Includes | Ground socket |
Shape | Conical |
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Height (Metric) | Socket Joint | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Height (Metric) | Socket Joint | Price | Quantity & Availability | ||
12599205
|
Quickfit
FE150/4 |
74 mm | 150 mL | 116 mm | 29/32 |
159.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
12692052
|
Quickfit
FE50/2 |
51 mm | 50 mL | 85 mm | 19/26 |
123.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10600701
|
Quickfit
FE25/1 |
42 mm | 25 mL | 70 mm | 14/23 |
143.00 €
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
10253540
|
Quickfit
FE150/2 |
74 mm | 150 mL | 118 mm | 19/26 |
159.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10345340
|
Quickfit
FE50/3 |
51 mm | 50 mL | 90 mm | 24/29 |
123.00 €
Pack of 5 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
10840861
|
Quickfit
FE250/3 |
85 mm | 250 mL | 130 mm | 24/29 |
194.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||
10243880
|
Quickfit
FE100/2 |
64 mm | 100 mL | 100 mm | 19/26 |
174.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
12081682
|
Quickfit
FE250/4 |
85 mm | 250 mL | 130 mm | 29/32 |
150.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10641861
|
Quickfit
FE25/2 |
42 mm | 25 mL | 70 mm | 19/26 |
145.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10508980
|
Quickfit
FE100/3 |
64 mm | 100 mL | 100 mm | 24/29 |
191.00 €
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10243400
|
Quickfit
FE1L/3 |
130 mm | 1000 mL | 220 mm | 24/29 |
75.40 €
Pack of 2 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
10743961
|
Quickfit
FE150/3 |
74 mm | 150 mL | 118 mm | 24/29 |
156.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10315060
|
Quickfit
FE100/1 |
64 mm | 100 mL | 100 mm | 14/23 |
135.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10263880
|
Quickfit
FE500/4 |
101 mm | 500 mL | 175 mm | 29/32 |
152.00 €
Pack of 5 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
10651861
|
Quickfit
FE500/3 |
101 mm | 500 mL | 175 mm | 24/29 |
155.00 €
Pack of 5 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
10851021
|
Quickfit
FE250/5 |
85 mm | 250 mL | 130 mm | 34/35 |
153.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||
10496840
|
Quickfit
FE50/1 |
51 mm | 50 mL | 85 mm | 14/23 |
123.00 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
12569205
|
Quickfit
FE100/4 |
64 mm | 100 mL | 100 mm | 29/32 |
135.00 €
Pack of 5 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||
12478916
|
Quickfit
FE250/2 |
85 mm | 250 mL | 130 mm | 19/26 |
187.00 €
Pack of 5 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
15577820
|
Quickfit
FE1L/4 |
130 mm | 1000 mL | 220 mm | 29/32 |
371.00 €
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||
15597820
|
Quickfit
FE2L/4 |
160 mm | 2000 mL | 270 mm | 29/32 |
404.00 €
Pack of 5 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||
15527830
|
Quickfit
FE500/2 |
101 mm | 500 mL | 175 mm | 19/26 |
610.00 €
Pack of 20 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||
15507830
|
Quickfit
FE2L/5 |
160 mm | 2000 mL | 270 mm | 34/35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15517830
|
Quickfit
FE5/1 |
25 mm | 5 mL | 54 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15587820
|
Quickfit
FE1L/5 |
130 mm | 1000 mL | 220 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15537830
|
Quickfit
FE500/5 |
101 mm | 500 mL | 175 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
Specifications
ISO 4797 | |
Borosilicate Glass | |
Ground socket | |
Conical |