missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Vacuum Filter Flask with Side Arm And Ground Socket

Filter flasks with precision-ground Quickfit™ socket.

Brand:  Quickfit FB1L/4

Code : NEW

Additional Details : Weight : 1.57000kg

 View more versions of this product

Product Code. 15547810

  • 635.00 € / Pack of 10


Call for latest local availability
01 885 5854
Add to basket

Description

Description

  • Complies with ISO 6556
  • With traditional plain glass side-arm to accept 9mm bore vacuum tubing
  • Heavy-wall and base construction for vacuum work
Specifications

Specifications

ISO 3585, DIN 12217, ASTM E-438
136 mm
Borosilicate Glass
Style: Erlenmeyer
1000 mL
227 mm
Videos
Documents