Learn More
Quickfit™ Borosilicate Glass Vacuum Filter Flask with Side Arm And Ground Socket
Filter flasks with precision-ground Quickfit™ socket.
725.00 € - 985.00 €
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | ||||
15547810
|
Quickfit
FB1L/4 |
1000 mL | 24/29 |
985.00 €
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
15567810
|
Quickfit
FB250/4 |
250 mL | 29/32 |
725.00 €
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
15587810
|
Quickfit
FB500/4 |
500 mL | 29/32 |
765.00 €
Pack of 10 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
Description
- Complies with ISO 6556
- With traditional plain glass side-arm to accept 9mm bore vacuum tubing
- Heavy-wall and base construction for vacuum work
Specifications
Borosilicate Glass |