Learn More
Quickfit™ Glass Joint Sockets
Manufactured with minimum shank length of 100mm
62.30 € - 386.00 €
Product Code | Brand | Joint Size | Thickness (Metric) Wall | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Thickness (Metric) Wall | Quantity | Price | Quantity & Availability | |||
10243270
|
Quickfit
SRB24UB |
24/29 | 2mm | 10/Pk. |
77.70 €
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
15577980
|
Quickfit
SRB60UB |
60/46 | 3.2mm | 1/Pk. |
386.00 €
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
10273700
|
Quickfit
SRB10UB |
10/19 | 1.5mm | 10/Pk. |
71.20 €
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
12989363
|
Quickfit
SRB34UB |
34/35 | 2mm | 10/Pk. |
147.00 €
Pack of 10 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
12999363
|
Quickfit
SRB40UB |
40/38 | 2.3mm | 5/Pk. |
94.20 €
Pack of 5 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
12425406
|
Quickfit
SRB19UB |
19/26 | 2mm | 10/Pk. |
77.70 €
Pack of 10 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||
12455406
|
Quickfit
SRB29UB |
29/32 | 2mm | 10/Pk. |
112.00 €
Pack of 10 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||
12485406
|
Quickfit
SRB45UB |
45/40 | 2.5mm | 5/Pk. |
99.90 €
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
10568790
|
Quickfit
SRB14UB |
14/23 | 1.8mm | 10/Pk. |
84.40 €
Pack of 10 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||
12603072
|
Quickfit
SRB7UB |
7/16 | 1.5mm | 10/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15557980
|
Quickfit
SRB12UB |
12/21 | 1.8mm | 10/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Manufactured with minimum shank length of 100mm
- Unbadged ground glass sockets
- Free from any ceramic markings
- Avoids problem of badge charring at high temperatures that can give untidy appearance
- High quality joints manufactured to accuracy that surpasses requirements of ISO 383
- Accurate taper and fine surface finish reduce possibility of joint seizure or leakage
- With round profile bead on top of socket for extra mechanical strength