missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Stirrer Guides
Designed to reduce excessive whip in glass stirrer shafts which can lead to breakage.
202.00 € - 494.00 €
Specifications
For Use With (Application) | To reduce excessive 'whip' in Glass stirrer shafts which can lead to breakage |
---|
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15547990
|
Quickfit
ST2/33C/SC |
494.00 €
Pack of 5 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||||
15537990
|
Quickfit
ST2/2/SC |
Cone size: 19/26; Length: 200mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15557990
|
Quickfit
ST2/44C/SC |
Cone size: 29/32; Length: 300mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- PTFE bearing is fitted to the bottom of guides ST2/33C/SC and ST2/44C/SC for additional support
- Features a side-arm with detachable plastic connector for the introduction of gases or connection to a vacuum supply
Specifications
To reduce excessive 'whip' in Glass stirrer shafts which can lead to breakage |