missing translation for 'onlineSavingsMsg'
Learn More

Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask

Manufactured from borosilicate glass. Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask's wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.

Brand:  Quickfit FR6LF

Code : NEW

Additional Details : Weight : 1.73000kg

 View more versions of this product

Product Code. 15547900

  • 875.00 € / Pack of 5

Estimated Shipment date
from Supplier 18-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • With large diameter Pyrex Labware™ Quickfit™ flat-flange joint
  • For use with Pyrex Labware™ Quickfit™ flat-flange lids and retaining clips
  • Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues
Specifications

Specifications

Spherical
6 L
Quickfit™ flat flange lids and retaining clips
Reaction flask
236 mm
Borosilicate Glass, Pyrex Glass
260 mm
Spherical
Videos
Documents