missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 97%

Brand:  Alfa Aesar™ H62328.MD

Code : NEW

Additional Details : CAS Number : 214852-61-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15475555

  • 100.00 € / 250mg


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Specifications

Chemical Identifiers

214852-61-6
533.71
CYMIEBREXUYHGN-BTQNPOSSSA-N
49853424
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
C29H47N3O6
MFCD01318750
z-d-dab boc-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, 2r-4-1,1-dimethylethyl oxy carbonyl amino-2-phenylmethyl oxy carbonyl amino butanoic acid dicyclohexylamine salt, z-d-dab boc-oh*dcha, n-a-z-n-?-boc-d-2,4-diaminobutyric aciddicyclohexylaminesalt, n-cbz-n'-boc-d-2,4-diaminobutyric acid dicyclohexylamine salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, 2r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha, 2r-4-1,1-dimethylethyl oxy carbonyl amino-2-phenylmethyl oxy carbonyl amino butanoic acid dicyclohexylamine salt;
N-cyclohexylcyclohexanamine;(2R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

(R)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
214852-61-6
MFCD01318750
CYMIEBREXUYHGN-BTQNPOSSSA-N
N-cyclohexylcyclohexanamine;(2R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)butanoic acid
49853424
Solid
250mg
C29H47N3O6
z-d-dab boc-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, 2r-4-1,1-dimethylethyl oxy carbonyl amino-2-phenylmethyl oxy carbonyl amino butanoic acid dicyclohexylamine salt, z-d-dab boc-oh*dcha, n-a-z-n-?-boc-d-2,4-diaminobutyric aciddicyclohexylaminesalt, n-cbz-n'-boc-d-2,4-diaminobutyric acid dicyclohexylamine salt, dicyclohexylamine r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoate, 2r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid-n-cyclohexylcyclohexanamine 1/1, 2r-2-benzyloxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid; dicha, 2r-4-1,1-dimethylethyl oxy carbonyl amino-2-phenylmethyl oxy carbonyl amino butanoic acid dicyclohexylamine salt;
CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
533.71
553.7
97%
Videos
SDS