missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylbutyric acid, 99%, ACROS Organics™

Brand:  Acros Organics 378520010

Code : 29

Additional Details : CAS Number : 938-79-4 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10025842

  • 99.76 € / 1g

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

938-79-4
164.2
OFJWFSNDPCAWDK-SECBINFHSA-N
785330
CCC(C1=CC=CC=C1)C(=O)O
C10H12O2
MFCD00063165
r---2-phenylbutyric acid, 2r-2-phenylbutanoic acid, 2-phenylbutyric acid, r, r-2-phenylbutyric acid, benzeneacetic acid, alpha-ethyl-, r, r-2-phenylbutanoic acid, 2r-2-phenyl-butanoic acid, 2r-2-phenyl-mutanoic acid, r---2-phenylbutanoic acid, benzeneacetic acid,a-ethyl-, ar
(2R)-2-phenylbutanoic acid

Specifications

(R)-(-)-2-Phenylbutyric acid
110°C
Glass Bottle
1.055
Colorless
938-79-4
100.0
C10H12O2
MFCD00063165
r---2-phenylbutyric acid, 2r-2-phenylbutanoic acid, 2-phenylbutyric acid, r, r-2-phenylbutyric acid, benzeneacetic acid, alpha-ethyl-, r, r-2-phenylbutanoic acid, 2r-2-phenyl-butanoic acid, 2r-2-phenyl-mutanoic acid, r---2-phenylbutanoic acid, benzeneacetic acid,a-ethyl-, ar
OFJWFSNDPCAWDK-SECBINFHSA-N
(2R)-2-phenylbutanoic acid
785330
Liquid
1.0550g/mL
Authentic
1.5150 to 1.5170 (20°C, 589nm)
130°C (3.0 mbar)
1g
98.5
98.5% min. (GC)
C2H5CH(C6H5)CO2H
09,IV,1815
Solubility in water: insoluble
CCC(C1=CC=CC=C1)C(=O)O
164.2
164.2
99%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (R)-(-)-2-Phenylbutyric acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents