Learn More
(R)-(+)-3-(Dimethylamino)pyrrolidine, 98%, ACROS Organics™
Brand: Acros Organics 426990010
Code : 29
Additional Details : CAS Number : 132958-72-6 Weight : 0.05100kg Transport : UN number : 2733 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
132958-72-6 | |
114.19 | |
AVAWMINJNRAQFS-ZCFIWIBFSA-N | |
2758521 | |
CN(C)C1CCNC1 |
C6H14N2 | |
MFCD00191347 | |
r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine | |
(3R)-N,N-dimethylpyrrolidin-3-amine |
Specifications
(R)-(+)-3-(Dimethylamino)pyrrolidine | |
Glass bottle | |
1g | |
97.5 | |
97.5% min. (GC) | |
MFCD00191347 | |
AVAWMINJNRAQFS-ZCFIWIBFSA-N | |
(3R)-N,N-dimethylpyrrolidin-3-amine | |
2758521 | |
98% |
51.7°C | |
166°C | |
132958-72-6 | |
100.0 | |
C6H14N2 | |
r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine | |
CN(C)C1CCNC1 | |
114.19 | |
114.19 |
Safety and Handling
- (R)-(+)-3-(Dimethylamino)pyrrolidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.