missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Pan Caspase Inhibitor Z-VKD-FMK Biotinylated

Product Code. 18684749
Click to view available options
Quantity:
0.5 mg
Unit Size:
500µg
This item is not returnable. View return policy

Product Code. 18684749

Brand: R&D Systems™ FMK011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an intracellular detector of apoptotic cells. R&D Systems Caspase-13 Inhibitor Z-LEED-FMK binds to the active site of activated proteases irreversibly and inhibits apoptosis.

  • Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying caspase activity
  • Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
  • Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Conjugate Biotin
Quantity 0.5 mg
Product Type Pan Caspase
Molecular Weight (g/mol) 863Da
Inhibitors Z-VKD-FMK
Form Lyophilized

For research use only. Not for use in diagnostic procedures

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.