Learn More
R&D Systems™ Human IL-28 R alpha/IFN-lambda R1 NP_734464 VersaClone cDNA
Dopamine Transporter
Brand: R&D Systems™ RDC1082
Additional Details : Weight : 0.00970kg
Description
The Human IL-28 R alpha/IFN-lambda R1 NP_734464 VersaClone cDNA clone contains the complete ORF for SLC6A3/DAT1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC6A3/DAT1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans