missing translation for 'onlineSavingsMsg'
Learn More

(R)-alpha-Methylaspartic acid, 98%, 98% ee, Thermo Scientific™

Catalog Number 11975351
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
14603-76-0
147.13
CWAYDJFPMMUKOI-RXMQYKEDSA-N
2-methyl-d-aspartic acid, alpha-methyl-d-asp, r---2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl, 2r-2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl-, hydrochloride 1:1, ambotzhaa7540, a-methyl-d-asp, h-a-me-d-asp-oh, r-2-methylaspartic acid
6993662
(2R)-2-amino-2-methylbutanedioic acid
CC(CC(=O)O)(C(=O)O)N
This item is not returnable. View return policy
14603-76-0
147.13
CWAYDJFPMMUKOI-RXMQYKEDSA-N
2-methyl-d-aspartic acid, alpha-methyl-d-asp, r---2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl, 2r-2-amino-2-methylbutanedioic acid, d-aspartic acid,2-methyl-, hydrochloride 1:1, ambotzhaa7540, a-methyl-d-asp, h-a-me-d-asp-oh, r-2-methylaspartic acid
6993662
(2R)-2-amino-2-methylbutanedioic acid
CC(CC(=O)O)(C(=O)O)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >249°C
Color White
Enantiomeric Excess > 75%
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Molecular Formula C5H9NO4
Linear Formula HOOCC(CH3)NH2CH2COOH
Quantity 500 mg
Specific Rotation -54
Formula Weight 147.13
Percent Purity 98%,98% ee
Physical Form Fine Crystalline Powder
Chemical Name or Material (R)-α-Methylaspartic acid, 99+%, (80% e.e.)
Show More Show Less
Product Identifier
  • (R)-alpha-Methylaspartic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'