missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-α-Methyltyrosine, 98%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | >211°C |
Color | White to Yellow |
Enantiomeric Excess | 98.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 1 g |
Specific Rotation | -18 |
Formula Weight | 195.22 |
Show More |
Safety and Handling
Product Identifier
- (R)-alpha-Methyltyrosine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'