Learn More
FZD9 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FZD9.
Brand: Abnova PAB16384.50ug
Additional Details : Weight : 0.02000kg
Description
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney. [provided by RefSeq]
Specifications
FZD9 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of FZD9. | |
In PBS (0.09% sodium azide) | |
O00144 | |
FZD9 | |
A synthetic peptide (conjugated with KLH) corresponding to human FZD9. | |
50 μg | |
Primary | |
3rd extracellular domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
FZD9 | |
CD349/FZD3 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
8326 | |
Human, Mouse, Rat | |
Liquid |
For Research Use Only