Learn More
Invitrogen™ Rabbit anti-Llama IgG (H+L) Secondary Antibody, Biotin
Rabbit Polyclonal Secondary Antibody
Brand:  Invitrogen™ A16158
Product Code. 15323956
  • 293.00 € / 1mg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Llama secondary antibodies are affinity-purified antibodies with well-characterized specificity for llama immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Llama IgG (H+L)
Polyclonal
Biotin
Rabbit
Affinity chromatography
RUO
Camelid
Secondary Antibody
IgG
Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
2.5 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.2
Gamma Immunoglobins Heavy and Light chains.
1 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.