Learn More
NCR3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NCR3.
Brand: Abnova PAB17794.100ug
Additional Details : Weight : 0.02000kg
Description
Natural cytotoxicity receptors (NCRs), such as NCR3, are activating natural killer (NK) cell receptors that belong to the immunoglobulin (Ig) superfamily. NCR3 is expressed in all resting and activated NK cells and forms a complex with CD3-zeta (CD3Z, or CD247; MIM 186780) (Sato et al., 2001 [PubMed 11782277]).[supplied by OMIM
Specifications
NCR3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NCR3. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
1C7/CD337/LY117/MALS/NKp30 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total NCR3 protein | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
NCR3 | |
NCR3 | |
A synthetic peptide corresponding to internal of human NCR3. | |
RUO | |
259197 | |
Human, Rat | |
Liquid |
For Research Use Only