missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Rainin™ Bioclean™ Pipette Tip, Packaged in Racks
POA
Specifications
For Use With (Application) | Perfect production and advanced design |
---|---|
Format | Rack |
For Use With (Equipment) | Rainin |
Specifications
Perfect production and advanced design | |
Rack | |
Rainin |