Learn More
Invitrogen™ RAT TWEAK R ELISA Kit
Rat TWEAK R quantitates rat TWEAK R in serum, plasma, supernatant.
Brand: Invitrogen™ ERA58RBX5
Additional Details : Weight : 0.33800kg
Description
Rat TWEAK R quantitates rat TWEAK R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat TWEAK R.
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.Specifications
AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12 A; Tnfrsf12a; Tumor necrosis factor receptor superfamily member 12 A; tumor necrosis factor receptor superfamily, member 12 A; TweakR; TWEAK-R; tweak-receptor; type I transmembrane protein Fn14 | |
0.3 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
CD266 | |
4 hr 45 min |
0.3 to 80 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
TNFRSF12A | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |