Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06008285
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11502234
  • 987.92 € / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Without graduation
  • With laboratory flange LF
  • With groove
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
6 L
385 mm
236 mm
Duran Glass
Lab Flange 100 LF