missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Description
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- UV protection due to external colouration
- All glass components
- Tight closure due to standard ground joint
Specifications
Capacity (Metric) | 1000 mL |
For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules |
Includes | With standard ground glass flat-headed stopper |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Diameter (Metric) | 110 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'