Learn More
Fisherbrand™ Recirculating Chiller for Sonicators Accessories: Tubing/Connector Set
Accessories for recirculating chiller
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.45000kg
Product Code. 15171293
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Tubing and connector set for use with Qsonica Recirculating Chiller for Sonicators.
Tubing/Connector Set for Cup Horn
  • Quick-connect tubing and fittings (sold separately) are used to attach Chiller (Cat. No. 15-338-421) to ports on the cup horn
  • Allows cold water to recirculate through the ultrasonic horn reservoir
Specifications
Specifications
Tubing and Connector Set
Ideal for attach chiller to ports on the cup horn
Q500 and Q700 Sonicator™
2 male fittings, 2 right angle male fittings and 2 female fittings