missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine ApoAI Protein
A DNA sequence encoding the Bos taurus (Bovine) ApoAI, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_776667.2 |
---|---|
Gene ID (Entrez) | 281631 |
Name | ApoAI Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097381
|
enQuireBio™
QP8986-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037391
|
enQuireBio™
QP8986-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007391
|
enQuireBio™
QP8986-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027391
|
enQuireBio™
QP8986-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047391
|
enQuireBio™
QP8986-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017391
|
enQuireBio™
QP8986-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776667.2 | |
ApoAI Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281631 | |
Research Use Only | |
APOA1 | |
Recombinant Protein | |
Yeast | |
DDPQSSWDRV KDFATVYVEA IKDSGRDYVA QFEASALGKQ LNLKLLDNWD TLASTLSKVR EQLGPVTQEF WDNLEKETAS LRQEMHKDLE EVKQKVQPYL DEFQKKWHEE VEIYRQKVAP LGEEFREGAR QKVQELQDKL SPLAQELRDR ARAHVETLRQ QLAPYSDDLR QRLTARLEAL KEGGGSLAEY HAKASEQLKA LGEKAKPVLE DLRQGLLPVL ESLKVSILAA IDEASKKLNA Q | |
Greater than 90% as determined by SDS-PAGE. |