missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine CD8A / MAL Protein
A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_776440.1 |
---|---|
Gene ID (Entrez) | 281060 |
Name | CD8A / MAL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987859
|
enQuireBio™
QP5800-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927869
|
enQuireBio™
QP5800-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997859
|
enQuireBio™
QP5800-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15917869
|
enQuireBio™
QP5800-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937869
|
enQuireBio™
QP5800-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15907869
|
enQuireBio™
QP5800-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776440.1 | |
CD8A / MAL Protein | |
Not determined. | |
Extracellular Domain | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281060 | |
Research Use Only | |
CD8A | |
Recombinant Protein | |
Yeast | |
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN | |
Greater than 90% as determined by SDS-PAGE. |