missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine CD8A / MAL Protein

A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5800-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987859

Quantity Price
1 914.06 € / 100µg
4 859.86 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_776440.1
CD8A / MAL Protein
100 μg
Not determined.
Extracellular Domain
Bovine
His
Tris-based buffer, 50% glycerol
281060
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD8A
Recombinant Protein
Yeast
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN
Videos
SDS
Documents
Certificates