missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine DNase Protein
A cDNA sequence encoding the DNase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11693-1gram
Additional Details : Weight : 0.01000kg
Specifications
282217 | |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Bovine | |
MRGTRLMGLL LALAGLLQLG LSLKIAAFNI RTFGETKMSN ATLASYIVRI VRRYDIVLIQ EVRDSHLVAV GKLLDYLNQD DPNTYHYVVS EPLGRNSYKE RYLFLFRPNK VSVLDTYQYD DGCESCGNDS FSREPAVVKF SSHSTKVKEF AIVALHSAPS DAVAEINSLY DVYLDVQQKW HLNDVMLMGD FNADCSYVTS SQWSSIRLRT SSTFQWLIPD SADTTATSTN CAYDRIVVAG SLLQSSVVPG SAAPFDFQAA YGLSNEMALA ISDHYPVEVT LT |
DNase Protein | |
Research Use Only | |
DNASE1 | |
Bovine | |
Untagged |