missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Trypsin Protein
A cDNA sequence encoding the Trypsin was constructed and used to recombinantly synthesize the protein.
131.00 € - 1015.00 €
Specifications
Name | Trypsin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | 3,650 Units/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975498
|
enQuireBio™
QP10896-1MG |
1 mg |
131.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965498
|
enQuireBio™
QP10896-10MG |
10 mg |
203.00 €
10mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955498
|
enQuireBio™
QP10896-100MG |
100 mg |
1015.00 €
100mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Trypsin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Bovine | |
The protein was lyophilized without any additives. |
Research Use Only | |
3,650 Units/mg. | |
Bovine | |
Untagged | |
Greater than 90% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |