missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis Large cysteine-rich periplasmic protein OmcB Protein

A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) Large cysteine-rich periplasmic protein OmcB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7015-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047050

  • 1440.00 € / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_219955.1
Large cysteine-rich periplasmic protein OmcB Protein
Research Use Only
omcB
Recombinant Protein
E. coli
LADTKAKDNT SHKSKKARKN HSKETPVDRK EVAPVHESKA TGPKQDSCFG RMYTVKVNDD RNVEITQAVP EYATVGSPYP IEITATGKRD CVDVIITQQL PCEAEFVRSD PATTPTADGK LVWKIDRLGQ GEKSKITVWV KPLKEGCCFT AATVCA
Greater than 90% as determined by SDS-PAGE.
884223
500 μg
Not determined.
Partial (see sequence information for more details).
C. trachomatis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions