missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine IFNG Protein

A cDNA sequence encoding the IFNG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12378-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990219

Quantity Price
1 171.64 € / 25µg
10 161.27 € / 25µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

403801
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
IFNG
Canine
His
IFNG protein solution (0.5 mg/ml) contains 20mM MES (pH6.0), 20% glycerol, 0.1M NaCl and 1mM EDTA.
IFNG Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQAMFFKE IENLKEYFNA SNPDVSDGGS LFVDILKKWR EESDKTIIQS QIVSFYLKLF DNFKDNQIIQ RSMDTIKEDM LGKFLNSSTS KREDFLKLIQ IPVNDLQVQR KAINELIKVM NDLSPRSNLR KRKRSQNLFR GRRASK
Videos
SDS
Documents
Certificates