missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine IFNG Protein

A cDNA sequence encoding the IFNG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10677-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984618

  • 3401.86 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

403801
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IFNG
Recombinant Protein
E. coli
QAMFFKEIEN LKEYFNASNP DVSDGGSLFV DILKKWREES DKTIIQSQIV SFYLKLFDNF KDNQIIQRSM DTIKEDMLGK FLNSSTSKRE DFLKLIQIPV NDLQVQRKAI NELIKVMNDL SPRSNLRKRK RSQNLFRGRR ASK
IFNG Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by an anti-viral assay using A-72 canine fibroma cells infected with vesicular stomatitis virus (VSV) is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Canine
Untagged
The IFNG was lyophilized from a 0.2microm filtered concentrated solution in 25mM Sodium Succinate, pH 5.0, 60mM NaCl, with 0.1% Tween-80.
Videos
SDS
Documents
Certificates