missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine IL 3 Protein

A cDNA sequence encoding the IL 3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10713-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984938

  • 161.30 € / 10µg

Estimated Shipment: 10-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

481497
Greater than 97.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IL3
Canine
Untagged
Lyophilized from a concentrated (1 mg/ml) solution in water containing 1X PBS pH 7.4.
IL 3 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
RPFSTDLPKQ YFTMINEIME MLNKSPSPSE EPLDSNEKET LLEDTLLRPN LDVFLNASSK FHKNGLLIWN NLKEFLPLLP TPTPRGEPIS IMENNWGDFQ RKLKKYLEAL DNFLNFKNKP
Videos
SDS
Documents
Certificates